Product Details

SNP ID
rs1229977
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:99281257 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACTGCTGGACCTTCTAGAAAATCT[C/T]GGCAATGGAAACGTGTTGGTCTCTG
Phenotype
MIM: 103700
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADH1A PubMed Links

Gene Details

Gene
ADH1A
Gene Name
alcohol dehydrogenase 1A (class I), alpha polypeptide
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000667.3 Intron NP_000658.1
Gene
LOC100507053
Gene Name
uncharacterized LOC100507053
There are no transcripts associated with this gene.

View Full Product Details