Product Details

SNP ID
rs5800
Assay Type
Functionally tested
NCBI dbSNP Submissions
51
Location
Chr.1:41479019 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCCAGCCTCAGGGGAGCCATCCAG[C/T]CTCCAGGGCTGGCTGAGGCATGCGT
Phenotype
MIM: 131241
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EDN2 PubMed Links

Gene Details

Gene
EDN2
Gene Name
endothelin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001302269.1 1009 UTR 3 NP_001289198.1
NM_001956.4 1009 UTR 3 NP_001947.1
XM_017000512.1 1009 UTR 3 XP_016856001.1

View Full Product Details