Product Details

SNP ID
rs13280283
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:98123981 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTTGTAACCCCAATATTAATACCC[A/G]TGAAGCTTTGGCTATCATTTGTGGA
Phenotype
MIM: 602486 MIM: 602487
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
POP1 PubMed Links

Gene Details

Gene
POP1
Gene Name
POP1 homolog, ribonuclease P/MRP subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145860.1 Intron NP_001139332.1
NM_001145861.1 Intron NP_001139333.1
NM_015029.2 Intron NP_055844.2
XM_011516801.2 Intron XP_011515103.1
Gene
RIDA
Gene Name
reactive intermediate imine deaminase A homolog
There are no transcripts associated with this gene.

View Full Product Details