Product Details

SNP ID
rs12419227
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:117144522 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCCGCGACCTCCCCTCCCCCACC[A/C]TTGCGGCCCCTCCCCCCACGCCGCC
Phenotype
MIM: 602508
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PAFAH1B2 PubMed Links

Gene Details

Gene
PAFAH1B2
Gene Name
platelet activating factor acetylhydrolase 1b catalytic subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184746.1 Intron NP_001171675.1
NM_001184747.1 Intron NP_001171676.1
NM_001184748.1 Intron NP_001171677.1
NM_001309431.1 Intron NP_001296360.1
NM_002572.3 Intron NP_002563.1
XM_017017840.1 Intron XP_016873329.1

View Full Product Details