Product Details

SNP ID
rs1123991
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:4681935 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAAGGCTGTTTCCAAAGCGGTGTAC[A/C]ACTGAGAGGCCAATAAGTGGCACAT
Phenotype
MIM: 611268
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
OR51E2 PubMed Links

Gene Details

Gene
OR51E2
Gene Name
olfactory receptor family 51 subfamily E member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030774.3 1021 Silent Mutation GTG,GTT V259V NP_110401.1

View Full Product Details