Product Details

SNP ID
rs12538847
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:34952764 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATGTGTCAACTTCCAAATACGGCA[C/G]AGTGAAGAAACTGGTAACTCCCATT
Phenotype
MIM: 613892
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DPY19L1 PubMed Links

Gene Details

Gene
DPY19L1
Gene Name
dpy-19 like 1 (C. elegans)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015283.1 Intron NP_056098.1
XM_011515245.2 Intron XP_011513547.1
XM_011515246.2 Intron XP_011513548.1

View Full Product Details