Product Details

SNP ID
rs6051628
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:3192033 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACCCATTGACAGAGACCCGCACAA[C/T]CATAAACCCTCTCATTCCCCTTCTT
Phenotype
MIM: 616177
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DDRGK1 PubMed Links

Gene Details

Gene
DDRGK1
Gene Name
DDRGK domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_023935.2 Intron NP_076424.1

View Full Product Details