Product Details

SNP ID
rs3850963
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:30631191 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAAAAAACGAGTGAAAAGAAAACAC[A/G]TCTCAGAGGTGGATTCTACTTCACT
Phenotype
MIM: 605600
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IPO8 PubMed Links

Gene Details

Gene
IPO8
Gene Name
importin 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190995.1 Intron NP_001177924.1
NM_006390.3 Intron NP_006381.2
XM_017018691.1 Intron XP_016874180.1
XM_017018692.1 Intron XP_016874181.1
XM_017018693.1 Intron XP_016874182.1

View Full Product Details