Product Details

SNP ID
rs4489629
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:216505939 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAACATTTTTAAAAATCTTCCTTCA[A/G]GATTCATTTGTCCTTCATTATTGTT
Phenotype
MIM: 602969
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ESRRG PubMed Links

Gene Details

Gene
ESRRG
Gene Name
estrogen related receptor gamma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134285.2 2746 UTR 3 NP_001127757.1
NM_001243505.1 2746 UTR 3 NP_001230434.1
NM_001243506.1 2746 UTR 3 NP_001230435.1
NM_001243507.1 2746 UTR 3 NP_001230436.1
NM_001243509.1 2746 UTR 3 NP_001230438.1
NM_001243510.2 2746 UTR 3 NP_001230439.1
NM_001243511.2 2746 UTR 3 NP_001230440.1
NM_001243512.1 2746 UTR 3 NP_001230441.1
NM_001243513.1 2746 UTR 3 NP_001230442.1
NM_001243514.1 2746 UTR 3 NP_001230443.1
NM_001243515.1 2746 UTR 3 NP_001230444.1
NM_001243518.1 2746 UTR 3 NP_001230447.1
NM_001243519.1 2746 UTR 3 NP_001230448.1
NM_001438.3 2746 UTR 3 NP_001429.2
NM_206594.2 2746 UTR 3 NP_996317.1
NM_206595.2 2746 UTR 3 NP_996318.1

View Full Product Details