Product Details
- SNP ID
-
rs2923241
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:62426847 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCTCAATGCAGACCAGATATCTGGT[A/G]GTCATGACGGACAAAGATCACACGA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
METTL2A
PubMed Links
Gene Details
- Gene
- METTL2A
- Gene Name
- methyltransferase like 2A
View Full Product Details