Product Details
- SNP ID
-
rs2853533
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.18:658064 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CGTTTAGTCCTAACCTCAATCCTGC[C/G]AGGGAGGGGACGCATCGTCCTCCTC
- Phenotype
-
MIM: 616990
MIM: 188350
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CLUL1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs77400748] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CLUL1
- Gene Name
- clusterin like 1
There are no transcripts associated with this gene.
- Gene
- TYMS
- Gene Name
- thymidylate synthetase
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001071.2 |
277 |
Intron |
|
|
NP_001062.1 |
- Gene
- TYMSOS
- Gene Name
- TYMS opposite strand
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001012716.2 |
277 |
Missense Mutation |
CGC,GGC |
R62G |
NP_001012734.2 |
View Full Product Details