Product Details

SNP ID
rs2242406
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:75540132 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGATAAGCAGGATGCTGACATACTG[C/T]ACCCAGGCGAACTTTACCATCTCCC
Phenotype
MIM: 604817 MIM: 614949
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CHST5 PubMed Links

Gene Details

Gene
CHST5
Gene Name
carbohydrate sulfotransferase 5
There are no transcripts associated with this gene.

Gene
TMEM231
Gene Name
transmembrane protein 231
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077416.2 987 Silent Mutation GTA,GTG V324V NP_001070884.2
NM_001077418.2 987 Silent Mutation GTA,GTG V271V NP_001070886.1

View Full Product Details