Product Details

SNP ID
rs4289995
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:111520791 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGAGATGGGAGCTGCACTCTGCA[C/T]CATTCAAAACAAATTAGCAGATAGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC00890 PubMed Links

Gene Details

Gene
LINC00890
Gene Name
long intergenic non-protein coding RNA 890
There are no transcripts associated with this gene.

View Full Product Details