Product Details

SNP ID
rs1803685
Assay Type
DME
NCBI dbSNP Submissions
1
Location
Chr.1:109737487 on Build GRCh38
Set Membership
DME Validated Inventoried
Context Sequence [VIC/FAM]
AACGGCACATGAAAGCCTTCAGGTT[C/T]GGGAACTCATCCAGGCACTTGGGGT
Phenotype
MIM: 138390
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GSTM3 PubMed Links

Gene Details

Gene
GSTM3
Gene Name
glutathione S-transferase mu 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000849.4 859 Silent Mutation CCA,CCG P183P NP_000840.2

View Full Product Details