Product Details
- SNP ID
-
rs3204790
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
29
- Location
-
Chr.1:12859209 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGGCCTTTCACCTGCCTCCCTCTGG[G/T]ATCGCTGATGAAGACGCTTCATCTG
- Phenotype
-
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
PRAMEF2
PubMed Links
Gene Details
- Gene
- PRAMEF2
- Gene Name
- PRAME family member 2
There are no transcripts associated with this gene.
View Full Product Details