Product Details

SNP ID
rs3204811
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:12860173 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAATGAACTCGAGGGATGGTTAGT[C/T]ACCAGATTCACCTCTGTGTTCCTCA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PRAMEF2 PubMed Links

Gene Details

Gene
PRAMEF2
Gene Name
PRAME family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_023014.1 Intron NP_075390.1
XM_011542004.1 Intron XP_011540306.1

View Full Product Details