Product Details

SNP ID
rs12610825
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:51795323 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGAACTGGGCACAGGAAAAGGATCT[A/C]AGCTGGTGGTAAGTTGGGGTCTGAA
Phenotype
MIM: 136539
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
FPR3 PubMed Links

Gene Details

Gene
FPR3
Gene Name
formyl peptide receptor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002030.4 178 UTR 5 NP_002021.3
XM_011526687.2 178 Intron XP_011524989.1

View Full Product Details