Product Details

SNP ID
rs12936959
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:41034287 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTCATGAATGGAACTGAAAGTGGA[A/T]ATTTCAAGTTGAAAATCAGCAAACT
Phenotype
MIM: 608819 MIM: 608820 MIM: 608821 MIM: 608822
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
KRTAP1-1 PubMed Links

Gene Details

Gene
KRTAP1-1
Gene Name
keratin associated protein 1-1
There are no transcripts associated with this gene.

Gene
KRTAP1-3
Gene Name
keratin associated protein 1-3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030966.1 569 UTR 3 NP_112228.1
Gene
KRTAP1-4
Gene Name
keratin associated protein 1-4
There are no transcripts associated with this gene.

Gene
KRTAP1-5
Gene Name
keratin associated protein 1-5
There are no transcripts associated with this gene.

View Full Product Details