Product Details
- SNP ID
-
rs2076988
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:142858366 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATCATATTTAGCATTTAATTGGAGC[A/C]CGATTAATTTTTTAAATTCTTACCA
- Phenotype
-
MIM: 602757
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
EPHB6
PubMed Links
Gene Details
- Gene
- EPHB6
- Gene Name
- EPH receptor B6
View Full Product Details