Product Details

SNP ID
rs547874515
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:175970538 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGATTCTCAGTATCTGAAATCAC[G/A]TAAATGTGATTCTTTGTATTTCTGC
Phenotype
MIM: 606929
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
THOC3 PubMed Links

Gene Details

Gene
THOC3
Gene Name
THO complex 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032361.3 Intron NP_115737.1
XM_017009985.1 Intron XP_016865474.1

View Full Product Details