Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001145795.1 | Intron | NP_001139267.1 | |||
NM_001145796.1 | Intron | NP_001139268.1 | |||
NM_001145797.1 | Intron | NP_001139269.1 | |||
NM_001145812.1 | Intron | NP_001139284.1 | |||
NM_001308293.1 | Intron | NP_001295222.1 | |||
NM_001308294.1 | Intron | NP_001295223.1 | |||
NM_015503.2 | Intron | NP_056318.2 | |||
XM_017023114.1 | Intron | XP_016878603.1 | |||
XM_017023115.1 | Intron | XP_016878604.1 | |||
XM_017023116.1 | Intron | XP_016878605.1 | |||
XM_017023117.1 | Intron | XP_016878606.1 | |||
XM_017023118.1 | Intron | XP_016878607.1 | |||
XM_017023119.1 | Intron | XP_016878608.1 | |||
XM_017023120.1 | Intron | XP_016878609.1 |