Product Details

SNP ID
rs4483492
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:15789749 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACAATGTTATTTAGTTTCTTGAA[A/G]GAATGTCAGTGATCCACTGATACTT
Phenotype
MIM: 611649
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM188A PubMed Links

Gene Details

Gene
FAM188A
Gene Name
family with sequence similarity 188 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318330.1 Intron NP_001305259.1
NM_024948.3 Intron NP_079224.1

View Full Product Details