Product Details
- SNP ID
-
rs953634
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:32731293 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGAAAAGTACCAAAAATGTGACTTT[A/C]TTAACCCTTGGGAAGAACTGAAATT
- Phenotype
-
MIM: 603850
MIM: 610957
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
DNM1L
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs138148657] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DNM1L
- Gene Name
- dynamin 1 like
- Gene
- YARS2
- Gene Name
- tyrosyl-tRNA synthetase 2
View Full Product Details