Product Details

SNP ID
rs2277548
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.15:78789766 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTTGATGCTGCAGCTGCGGTGCGG[C/T]TGGCACATGCCCGCCACATGGGACA
Phenotype
MIM: 605009
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
ADAMTS7 PubMed Links

Gene Details

Gene
ADAMTS7
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014272.4 1344 Silent Mutation CAA,CAG Q367Q NP_055087.2
XM_005254137.3 1344 Silent Mutation CAA,CAG Q367Q XP_005254194.1
XM_011521166.2 1344 Intron XP_011519468.1

View Full Product Details