Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001271989.1 | Intron | NP_001258918.1 | |||
NM_024899.3 | Intron | NP_079175.2 | |||
XM_005258149.4 | Intron | XP_005258206.1 | |||
XM_017025981.1 | Intron | XP_016881470.1 | |||
XM_017025982.1 | Intron | XP_016881471.1 | |||
XM_017025983.1 | Intron | XP_016881472.1 | |||
XM_017025984.1 | Intron | XP_016881473.1 | |||
XM_017025985.1 | Intron | XP_016881474.1 | |||
XM_017025986.1 | Intron | XP_016881475.1 | |||
XM_017025987.1 | Intron | XP_016881476.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_020232.4 | Intron | NP_064617.2 | |||
NM_147163.1 | Intron | NP_671692.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001128626.1 | Intron | NP_001122098.1 | |||
NM_001128627.1 | Intron | NP_001122099.1 | |||
NM_020148.2 | Intron | NP_064533.3 |