Product Details

SNP ID
rs4422957
Assay Type
Functionally tested
NCBI dbSNP Submissions
43
Location
Chr.1:74198465 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACTTCCCGTTTACTTCTCATCTG[C/T]AGGCCTATGACGCTCCCCAGGAGTA
Phenotype
MIM: 603609
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FPGT PubMed Links

Gene Details

Gene
FPGT
Gene Name
fucose-1-phosphate guanylyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199328.2 Intron NP_001186257.2
NM_001199329.2 Intron NP_001186258.2
NM_003838.4 Intron NP_003829.3
Gene
FPGT-TNNI3K
Gene Name
FPGT-TNNI3K readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001112808.2 Intron NP_001106279.2
NM_001199327.1 Intron NP_001186256.2
Gene
LRRIQ3
Gene Name
leucine rich repeats and IQ motif containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001105659.1 Intron NP_001099129.1
NM_001322315.1 Intron NP_001309244.1
XM_011540658.1 Intron XP_011538960.1
XM_011540659.1 Intron XP_011538961.1
XM_011540660.1 Intron XP_011538962.1
XM_011540662.2 Intron XP_011538964.1
XM_011540663.2 Intron XP_011538965.1
XM_017000286.1 Intron XP_016855775.1
XM_017000287.1 Intron XP_016855776.1
XM_017000288.1 Intron XP_016855777.1

View Full Product Details