Product Details

SNP ID
rs8134957
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:34499581 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGCTTCCCCGGGCTAAAGCTGGAG[A/C]ATGGGAATGTCTACAAGGCTCTTCC
Phenotype
MIM: 176261
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
KCNE1 PubMed Links

Gene Details

Gene
KCNE1
Gene Name
potassium voltage-gated channel subfamily E regulatory subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000219.5 Intron NP_000210.2
NM_001127668.3 Intron NP_001121140.1
NM_001127669.3 Intron NP_001121141.1
NM_001127670.3 Intron NP_001121142.1
NM_001270402.2 Intron NP_001257331.1
NM_001270403.2 Intron NP_001257332.1
NM_001270404.2 Intron NP_001257333.1
NM_001270405.2 Intron NP_001257334.1
XM_017028342.1 Intron XP_016883831.1

View Full Product Details