Product Details

SNP ID
rs2695316
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:177551579 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTGTGTCTTGCTTCCTGTACTTG[C/T]ATGGTAAGTTTCCGGAGGACCTCAG
Phenotype
MIM: 603051
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AGPS PubMed Links

Gene Details

Gene
AGPS
Gene Name
alkylglycerone phosphate synthase
There are no transcripts associated with this gene.

Gene
TTC30B
Gene Name
tetratricopeptide repeat domain 30B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152517.2 1218 Silent Mutation NP_689730.2

View Full Product Details