Product Details

SNP ID
rs2306900
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:28957348 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCCCTCCTACTCAATTGACACTTAC[A/G]GAGACATTTAGGGGCCCGGTCACTC
Phenotype
MIM: 616666
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PHF12 PubMed Links

Gene Details

Gene
PHF12
Gene Name
PHD finger protein 12
There are no transcripts associated with this gene.

Gene
SEZ6
Gene Name
seizure related 6 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098635.1 2688 Silent Mutation CTG,TTG L832L NP_001092105.1
NM_001290202.1 2688 Silent Mutation CTG,TTG L707L NP_001277131.1
NM_178860.4 2688 Silent Mutation CTG,TTG L832L NP_849191.3
XM_011524315.1 2688 Silent Mutation CTG,TTG L832L XP_011522617.1
XM_011524317.2 2688 Silent Mutation CTG,TTG L832L XP_011522619.1

View Full Product Details