Product Details

SNP ID
hCV26807155
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:70480862 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGAGGGCAGAGCATGGAGTGGGT[T/C]CAGACTTTGTTTCTCTGCTGCCAGC
Phenotype
MIM: 606976 MIM: 608675
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
COG4 PubMed Links

Gene Details

Gene
COG4
Gene Name
component of oligomeric golgi complex 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195139.1 2529 UTR 3 NP_001182068.1
NM_015386.2 2529 UTR 3 NP_056201.2
Gene
FUK
Gene Name
fucokinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145059.2 2529 Intron NP_659496.2

View Full Product Details