Product Details

SNP ID
rs1804600
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.16:30444923 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCTTTACTTTATTCCATCTTTCAG[G/C]ATTATCCAGCCATTGGTGGGCATTG
Phenotype
MIM: 606218
Polymorphism
G/C, Transversion Substitution
Allele Nomenclature
Literature Links
SEPHS2 PubMed Links

Gene Details

Gene
SEPHS2
Gene Name
selenophosphate synthetase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012248.3 1053 Missense Mutation CCT,GCT P269A NP_036380.2

View Full Product Details