Product Details

SNP ID
rs4490099
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:40225584 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CGTCAGCCACAGCTGCCTTCTTTTT[A/G]GCTCCTAGACTATACCAAGCCCATT
Phenotype
MIM: 164731
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AKT2 PubMed Links

Gene Details

Gene
AKT2
Gene Name
AKT serine/threonine kinase 2
There are no transcripts associated with this gene.

Gene
CNTD2
Gene Name
cyclin N-terminal domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024877.3 Intron NP_079153.2
XM_006723395.3 Intron XP_006723458.2
Gene
TTC9B
Gene Name
tetratricopeptide repeat domain 9B
There are no transcripts associated with this gene.

View Full Product Details