Product Details

SNP ID
rs1063464
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:72060473 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGAGTGATGCCCATCTGCCTACC[A/T]TCAAAGGATTATGCAGAAGTAGGGC
Phenotype
MIM: 140100 MIM: 140210
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
HP PubMed Links

Gene Details

Gene
HP
Gene Name
haptoglobin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001126102.2 688 Silent Mutation CCA,CCT P209P NP_001119574.1
NM_001318138.1 688 Silent Mutation CCA,CCT P209P NP_001305067.1
NM_005143.4 688 Silent Mutation CCA,CCT P268P NP_005134.1
Gene
HPR
Gene Name
haptoglobin-related protein
There are no transcripts associated with this gene.

Gene
TXNL4B
Gene Name
thioredoxin like 4B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142317.1 688 Intron NP_001135789.1
NM_001142318.1 688 Intron NP_001135790.1
NM_001324354.1 688 Intron NP_001311283.1
NM_001324355.1 688 Intron NP_001311284.1
NM_017853.2 688 Intron NP_060323.1
XM_017023377.1 688 Intron XP_016878866.1

View Full Product Details