Product Details

SNP ID
rs2708840
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:53493860 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGGGGTGAGAGCATTCTTCCATCA[C/T]GTGAGATCCTACAGGAAAATCATTC
Phenotype
MIM: 606043
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF331 PubMed Links

Gene Details

Gene
ZNF331
Gene Name
zinc finger protein 331
There are no transcripts associated with this gene.

Gene
ZNF813
Gene Name
zinc finger protein 813
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004301.3 Intron NP_001004301.2

View Full Product Details