Product Details

SNP ID
rs12741733
Assay Type
Functionally Tested
NCBI dbSNP Submissions
25
Location
Chr.1:246728370 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTCCACTGCTGCACTCTATAATGGA[C/T]GCCCTCATCTATCTCCCTTGCCTTT
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
SCCPDH PubMed Links

Gene Details

Gene
SCCPDH
Gene Name
saccharopine dehydrogenase (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016002.2 Intron NP_057086.2

View Full Product Details