Product Details

SNP ID
rs8027679
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:59638573 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAAGAAGGAAAGTTTTTTGGTTTT[C/G]TTTTTGCTATGATTGTCTAACTTTT
Phenotype
MIM: 600519
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GTF2A2 PubMed Links

Gene Details

Gene
GTF2A2
Gene Name
general transcription factor IIA subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320929.1 1293 UTR 3 NP_001307858.1
NM_001320930.1 1293 UTR 3 NP_001307859.1
NM_004492.2 1293 UTR 3 NP_004483.1

View Full Product Details