Product Details

SNP ID
rs11855587
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.15:68198619 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTATGAAAAGCTGTATGTGTTCTC[A/G]TTGTTGAAAATACAGGAAACAATAA
Phenotype
MIM: 606725 MIM: 603566
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
CALML4 PubMed Links

Gene Details

Gene
CALML4
Gene Name
calmodulin like 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031733.2 Intron NP_001026903.2
NM_001286694.1 Intron NP_001273623.1
NM_001286695.1 Intron NP_001273624.1
NM_033429.2 Intron NP_219501.2
Gene
CLN6
Gene Name
ceroid-lipofuscinosis, neuronal 6, late infantile, variant
There are no transcripts associated with this gene.

Gene
PIAS1
Gene Name
protein inhibitor of activated STAT 1
There are no transcripts associated with this gene.

View Full Product Details