Product Details
- SNP ID
-
rs2905542
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:36104696 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAGGCAAGGGTGAGGGCTGGATTTT[A/T]AAAGAGGGCCTGTTTTGGGGAGGGG
- Phenotype
-
MIM: 182284
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CCL4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs150004330] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CCL4
- Gene Name
- C-C motif chemokine ligand 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_002984.3 |
|
Intron |
|
|
NP_002975.1 |
View Full Product Details