Product Details

SNP ID
rs3912385
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:30744565 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAACTGAGAATATTCGCAGTGCAC[A/G]CACATTTTTCTTTACTAGAATTTGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SUZ12P1 PubMed Links

Gene Details

Gene
SUZ12P1
Gene Name
SUZ12 polycomb repressive complex 2 subunit pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details