Product Details

SNP ID
rs6469859
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:86110868 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGACAGGACAGGACAGGACAGGAC[C/T]GGCATCATAAGACCTCCGCCAGGCA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATP6V0D2 PubMed Links

Gene Details

Gene
ATP6V0D2
Gene Name
ATPase H+ transporting V0 subunit d2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152565.1 Intron NP_689778.1

View Full Product Details