Product Details

SNP ID
rs1984937
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:45903164 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCAAGAACGAAGTGCTGGGAAGGAC[A/C]AATTCATGGAACCGTGGGACGGTGC
Phenotype
MIM: 157140
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MAPT PubMed Links

Gene Details

Gene
MAPT
Gene Name
microtubule associated protein tau
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001123066.3 Intron NP_001116538.2
NM_001123067.3 Intron NP_001116539.1
NM_001203251.1 Intron NP_001190180.1
NM_001203252.1 Intron NP_001190181.1
NM_005910.5 Intron NP_005901.2
NM_016834.4 Intron NP_058518.1
NM_016835.4 Intron NP_058519.3
NM_016841.4 Intron NP_058525.1
XM_005257362.4 Intron XP_005257419.1
XM_005257364.4 Intron XP_005257421.1
XM_005257365.4 Intron XP_005257422.1
XM_005257366.3 Intron XP_005257423.1
XM_005257367.4 Intron XP_005257424.1
XM_005257368.4 Intron XP_005257425.1
XM_005257369.4 Intron XP_005257426.1
XM_005257370.4 Intron XP_005257427.1
XM_005257371.4 Intron XP_005257428.1
Gene
MAPT-AS1
Gene Name
MAPT antisense RNA 1
There are no transcripts associated with this gene.

Gene
MAPT-IT1
Gene Name
MAPT intronic transcript 1
There are no transcripts associated with this gene.

View Full Product Details