Product Details

SNP ID
rs2227823
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:76719083 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCCGCATGCCACTCCCTCTGGCC[A/G]GAATGCCCTCCCCATATCCCATACC
Phenotype
MIM: 187930
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
F2R PubMed Links

Gene Details

Gene
F2R
Gene Name
coagulation factor II thrombin receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001311313.1 Intron NP_001298242.1
NM_001992.4 Intron NP_001983.2
Gene
NCRUPAR
Gene Name
non-protein coding RNA, upstream of F2R/PAR1
There are no transcripts associated with this gene.

View Full Product Details