Product Details

SNP ID
rs4399068
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:27819465 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAAAAGAGAGTTCCAGAGATTTCCA[A/G]GTCCTGTCAGGAGTTAAGTTAAAGA
Phenotype
MIM: 300761
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MAGEB10 PubMed Links

Gene Details

Gene
MAGEB10
Gene Name
MAGE family member B10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182506.3 Intron NP_872312.2

View Full Product Details