Product Details

SNP ID
rs210587
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:152133937 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATAGAGTGAGACTTTGTTAAAAAAA[A/C]AAAAAGAAGAAGAAGAAAAAGAAAA
Phenotype
MIM: 300343
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MAGEA10 PubMed Links

Gene Details

Gene
MAGEA10
Gene Name
MAGE family member A10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001011543.2 2117 UTR 3 NP_001011543.2
NM_001251828.1 2117 UTR 3 NP_001238757.1
NM_021048.4 2117 UTR 3 NP_066386.2
Gene
MAGEA10-MAGEA5
Gene Name
MAGEA10-MAGEA5 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204811.2 2117 Intron NP_001191740.1

View Full Product Details