Product Details

SNP ID
rs3798352
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:34767032 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTTGTGCTTCATCTGAAAAAAAAA[T/A]TTTTAAATTGATTGCATTAGACTTT
Phenotype
MIM: 603522
Polymorphism
T/A, Transversion Substitution
Allele Nomenclature
Literature Links
SNRPC PubMed Links

Gene Details

Gene
SNRPC
Gene Name
small nuclear ribonucleoprotein polypeptide C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003093.2 Intron NP_003084.1

View Full Product Details