Product Details

SNP ID
rs2297225
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:19176987 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTGCGCTTGGTCTTGATGGTGGC[G/A]ATGGCCGCGTTGACATCTTTCGGGA
Phenotype
MIM: 602528
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
LOC101928697 PubMed Links

Gene Details

Gene
LOC101928697
Gene Name
uncharacterized LOC101928697
There are no transcripts associated with this gene.

Gene
TUBA3C
Gene Name
tubulin alpha 3c
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006001.2 Intron NP_005992.1

View Full Product Details