Product Details

SNP ID
rs2401149
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:42977171 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTCCATTTCCAGAGCACAGGCAA[A/G]AGTAGACGTAACATAATTCTTCAGA
Phenotype
MIM: 602100
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PKNOX1 PubMed Links

Gene Details

Gene
PKNOX1
Gene Name
PBX/knotted 1 homeobox 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286258.1 Intron NP_001273187.1
NM_001320694.1 Intron NP_001307623.1
NM_004571.4 Intron NP_004562.2

View Full Product Details