Product Details
- SNP ID
-
rs11677413
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:191953876 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TAGTGCCCAGTTACCTGTAGGGTGC[C/T]GCATTCATTTTTTTTTTTTTTTTTT
- Phenotype
-
MIM: 605734
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
TMEFF2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs563154787] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- TMEFF2
- Gene Name
- transmembrane protein with EGF like and two follistatin like domains 2
View Full Product Details