Product Details

SNP ID
rs4850244
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:131730654 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATATATTATTGTTACAACCAAAAG[A/C]AAACCTTGATAAACTCCAAATCTGA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
C2orf27A PubMed Links

Gene Details

Gene
C2orf27A
Gene Name
chromosome 2 open reading frame 27A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013310.3 Intron NP_037442.3

View Full Product Details