Product Details

SNP ID
rs744308
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:30397965 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACGAACTAAATCTCTTATCTGTC[A/C]CCCCTGGGGAGTGAGGAGGTCGGCT
Phenotype
MIM: 607558
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SEC14L2 PubMed Links

Gene Details

Gene
SEC14L2
Gene Name
SEC14 like lipid binding 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204204.2 Intron NP_001191133.1
NM_001291932.1 Intron NP_001278861.1
NM_012429.4 Intron NP_036561.1
NM_033382.2 Intron NP_203740.1

View Full Product Details